Learn More
Agencinox™ Flat Bottom Stainless Steel Dishes
Product Code. 11766533
Change view
Click to view available options
Capacity (Metric):
60 mL
100 mL
250 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11766533 250 mL Each
11756533 100 mL Each
11746533 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11766533 Supplier Agencinox™ Supplier No. 310
Use these dishes in decontamination services, operating rooms, surgeries, clinics and healthcare fields. Agencinox™ Flat Bottom Stainless Steel Dishes are ideal for holding dressings and other items which may be needed during surgical procedures.
  • Flat bottom stainless steel lotion bowls
  • Autoclavable
  • Available in a variety of sizes
Specifications
Bottom Shape Flat
Capacity (Metric) 250 mL
Material Stainless Steel
Shape Round
Type Lotion Bowl
Height (Metric) 50 mm
Diameter (Metric) 100 mm