Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck
Product Code. p-7222607
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
50 mL
100 mL
250 mL
1 L
2 L
Height (Metric):
90 mm
105 mm
138 mm
190 mm
230 mm
240 mm
missing translation for 'unitSize'
Pack of 10
Product Code. 10017839
missing translation for 'mfr': Lenz Laborglasintrumente 07050049
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 24450
  • With beaded rim
  • Material: DURAN™
Specifications
Capacity (Metric) 250 mL
Material Duran Glass
Height (Metric) 138 mm