Learn More
Benchmark Scientific™ Flat Block for Custom Drilling
Catalog Number 17944402
Click to view available options
Benchmark Scientific™ Flat Block
This flat block can be custom drilled for specific tubes sizes. Drilling charge extra. Fits Benchmark MultiTherm.
  • Flat block for custom drilling
  • Drilling charges extra
  • Fits MultiTherm H and HC Models
Description Flat block for custom drilling
Autoclavable Not Autoclavable
Chamber Material Aluminum
Holds Custom
Material Aluminum
No. of Places Custom
Type Block