Learn More
Grant Instruments™ Flask Platforms for use with ES-20/80
Product Code. 17765019
Click to view available options
Holds:
16 x 250 to 300 mL Flasks
30 x 100 to 150 mL Flasks
6 x 1000 mL Flasks
9 x 500 mL Flasks
Unit Size:
Each
Product Code. 17765019
Brand: Grant Instruments™ HSP6/1000
Only null left
Add to basket
Add to basket
The Grant™ interchangeable flask platforms are for use with the ES-20/80 Orbital Shaker-Incubator. The platforms have clamps to securely hold the flasks in position during vigorous mixing.
  • Interchangeable
  • Clamps securely hold flasks in place during vigorous mixing
  • Optional accessories for the ES-20/80 Orbital Shaker-Incubator
TRUSTED_SUSTAINABILITY
Specifications
Holds 6 x 1000 mL Flasks
Dimensions (L x W x H) 36 x 40 x 10 cm
Weight (English) 7.3 lb.
Weight (Metric) 3.3 kg
Description Platform for 1 L Flasks (6 x 1000 mL Flasks. ES-20/80 and ES-20/80 C)
Model HSP-6/1000
For Use With (Equipment) ES-20/80 and ES-20/80C