Learn More
Paragon Scientific™ Flash Point Certified Reference Material
Product Code. 15480464
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480464 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15480464 Supplier Paragon Scientific™ Supplier No. CRMUPMGO
  • ONLINE EXCLUSIVE PRICE
    3085.00 NOK
    2910.51 NOK / 250mL
    Save 174.49 NOK
    5% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Flash Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Flash point according to ASTM D93.
  • Certified Reference Material for annual verification checks in accordance with ASTM D93.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Product Type MCC Diesel UKAS ISO 17034 Certified (56.2 °C)
Certifications/Compliance Certified Reference Material for annual verification checks in accordance with ASTM D93
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • CRMU-PMGO
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
Nominal values quoted