Learn More
Macherey-Nagel™ Flash Chromatography Kit I
Product Code. 10515561 Shop All Macherey-Nagel Products
Change view
Click to view available options
Type:
Flash Chromatography Kit I
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10515561 Flash Chromatography Kit I Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10515561 Supplier Macherey-Nagel™ Supplier No. 727450
  • ONLINE EXCLUSIVE PRICE
    6625.00 NOK
    6271.60 NOK / Pack of 1
    Save 353.40 NOK
    5% Off
Estimated Shipment: 26-11-2026
Specifications
Type Flash Chromatography Kit I
Includes Includes One 450 x 20 mm I.D. Glass Column, One 1-L eluent Reservoir, 100 g Silica 60 (40 to 63 μm), Sea Sand, Silanized Glass Fiber Wadding, 1 m PTFE Tubing
For Use With (Application) Low Pressure Flash Chromatography