missing translation for 'onlineSavingsMsg'
Learn More

FKBP65 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16859723 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16859723 20 μL 20µL
16849723 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16859723 Supplier Proteintech Supplier No. 121721AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.

Specifications

Antigen FKBP65
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FKBP10
Gene Accession No. Q61576, Q96AY3
Gene Alias 65 kDa FK506 binding protein, 65 kDa FKBP, FK506 binding protein 10, FKBP 10, FKBP 65, FKBP10, FKBP6, FKBP65, hFKBP65, Immunophilin FKBP65, PPIase FKBP10, Rotamase
Gene Symbols Fkbp10
Host Species Rabbit
Immunogen FKBP65 Fusion Protein Ag2814
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14230, 360627, 60681
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.