Description
Maximum 2 per platform
Specifications
Specifications
| Holds | 40 x 15 mL Tubes |
| Positions | 40 |
| For Use With (Equipment) | Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker |
| Dimensions (L x W x H) | 262 x 112 x 140 mm |
| Diameter (Metric) Outer | 18 mm |
| Product Type | Fixed Tube Rack |