Learn More
BINDER™ Shelf, Reinforced
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Brand: BINDER™ 8012-0287
Additional Details : Weight : 2.00000kg
Description
- Reinforced
- Positioned at bottom level
- Maximum load of 45 kg
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system | |
BINDER Chambers KB 53 | |
Max. load of 67 kg | |
Shelf |