Description
- HILED modules provide imaging performance for fluorescence applications.
- Choose from six options ranging from UV to IR light.
- Each module has a narrow band pass filter to minimise cross-talk between different wavelengths making it ideal for multiplexing with fluorescent gels or blots.
- Syngene GBox chemi systems are all compatible with these modules.
- The new HILED modules offer reduced exposure time, an extended LED life, and a 20 x 20 cm viewing area.
Specifications
Specifications
| Product Line | Syngene Gel Doc |
| For Use With (Application) | Imaging DNA Gels and Multiplexing |
| Color | Blue |
| Product Type | HILEDS in Blue |