Learn More
Hettich Lab Technology™ Fixed Angle Rotor
Product Code. p-8009506
Change view
Click to view available options
Capacity:
12 x 1.1 to 15 mL
Max. Speed:
6,000 rpm
Max. RCF:
4,146 x g
Unit Size:
Each
Product Code. Capacity Max. Speed Max. RCF unitSize
10191372 12 x 1.1 to 15 mL 6,000 rpm 4,146 x g Each
1 options
Product Code. 10191372
Brand: Hettich Lab Technology™ 1613
Fixed Angle Rotor has a relatively low drag for high-speed centrifugation and oblique angling to speed up sedimentation.
  • Used primarily in the higher speed range.
  • Relatively low drag.
  • Oblique angling of tubes shortens the overall distance that particles must travel, thereby speeding up sedimentation.
Specifications
Type Fixed Angle Rotor
Capacity 12 x 1.1 to 15 mL
Max. RCF 4,146 x g
Max. Speed 6,000 rpm
Max. Tube Length 125 mm
Max. Tube Diameter 17 mm
For Use With (Equipment) Universal 320, Rotofix 32A
Angle 35°
No. of Places 12