Learn More
Invitrogen™ Fixation Medium (Medium A)
Product Code. 10572625
Change view
Click to view available options
Quantity:
5 mL
100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10572625 5 mL Each
10416103 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10572625 Supplier Invitrogen™ Supplier No. GAS001S5
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
Fixation Medium (Medium A)
Fixation Medium (Medium A) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.
The FIX & PERM™ Cell Permeabilization Kit is:
  • Flexible—compatible with multiple sample types and cellular antigens
  • Robust—scatter pattern of cells unaffected
  • Efficient—simultaneous detection of intracellular and surface markers
    View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
    The Fixation Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.
    How It Works
    Cells are incubated for 15 minutes at room temperature with the Fixation Medium and washed. Cells can then be incubated with Permeabilization Medium (Medium B) (Cat. No. GAS002S100) and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Content And Storage Store at room temperature.
    Quantity 5 mL
    Volume (Metric) 5 mL
    Reagent Type Fixation Reagent
    Shipping Condition Room Temperature
    Product Type Fixation Medium