Learn More
SCAT™ Hexagonal Installation Wrench for HPLC Systems
Shop All SCAT Europe Products
Click to view available options
Color:
Silver
Unit Size:
Each
Description
SCAT™ fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Specifications
| Type | Wrench |
| Color | Silver |
| For Use With (Application) | Solvent Handling |
| For Use With | PFA Fittings |
| Material | Aluminum |
| Autoclavable | Autoclavable |