Learn More
Hamilton™ Fitting with Washer
Description
- For gauges 18 to 22
- For thread 1/4 in.-28
- Sold in packs of 10
Specifications
Specifications
| Material | Metal (Washer) |
| Needle Gauge | 18 to 22 G |
| Quantity | 10/Pk. |