Learn More
BRAND™ Fitting for Dr. Schilling Pattern Automatic Burette
Product Code. 10340151 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
15 mL Dr. Schilling Pattern Automatic Burette
25 to 50 mL Dr. Schilling Pattern Automatic Burette
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10340151 15 mL Dr. Schilling Pattern Automatic Burette Each
10175640 25 to 50 mL Dr. Schilling Pattern Automatic Burette Each
2 options
Product Code. 10340151
Brand: BRAND™ 23746
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Micrometer screw with press button, including support clamp
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 15 mL
For Use With (Equipment) 15 mL Dr. Schilling Pattern Automatic Burette
Includes Micrometer Screw with Press Button, Support Clamp
Type Buret Fitting