Learn More
Fisherbrand™ Write-On Tape
Product Code. 17897291 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Width (English):
0.5 in.
1.5 in.
Length (English):
14 yd.
60 yd.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Length (English) Unit Size
17897291 1.5 in. 14 yd. Each
17807301 0.5 in. 60 yd. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17897291
Estimated Shipment: 21-12-2026
Sticks to glass, plastic, paper and wood.
  • Can be used in temperatures from -23°C to +121°C (-9°F to +250°F)
  • Well suited for use in autoclaves, refrigerators, deep freeze, sterilizers, and incubators
  • Easily marked with pencil, pen, ballpoint and most felt-tip markers
  • Removes without residue
  • Resists oil, water, and acid
Specifications
Color White
Format Tape
Product Type Write-On™ Tape
For Use With (Application) Autoclave, Refrigerator/freezer, Sterilizer, Incubator
Quantity 1 Ea.
Temperature Range (English) -9 to 250°F
Temperature Range (Metric) -23°C to +121°C
Length (English) 14 yd.
Width (English) 1.5 in.