Learn More
Fisherbrand™ Wide-Mouth Polypropylene Jars
Product Code. p-3217584 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Closure Size:
89 mm
Unit Size:
Pack of 6
Product Code. Capacity (Metric) Closure Size unitSize
11828661 250 mL 89 mm Pack of 6
1 options
Product Code. 11828661
Brand: Fisherbrand™ 0610140
Only null left
Add to basket
Add to basket
Includes
Polypropylene linerless screw closures
Higher temperature resistance than LDPE or HDPE.
  • Handle up to 135°C (275°F) for short periods and 100°C (212°F) continuously
  • Brittleness temperature of 0°C (32°F)
  • Rigid walls
  • Excellent chemical resistance
  • Can be sterilized by all normal methods, except dry heat and radiation
Completely disengage threads or remove cap before autoclaving.
Specifications
Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Natural
Description Jar PP Natural 8oz
Diameter (English) 3.5 in.
For Use With (Application) Storing Lab Specimen, Supplies or Powder
Includes Polypropylene Linerless Screw Closure
Material Polypropylene
Max. Temperature (Metric) 135°C
Max. Temperature (English) 275°F
No. per Case 36
No. per Pack 6
Quantity Case, Pack
Sterility Non-sterile
Closure Color White
Liner Material Linerless
Clarity Natural
Closure Included Yes
Graduated No
Mouth Wide
Shape Round
Height (Metric) 66 mm
Product Type Jar
Height (English) 2.5 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 89 mm
Diameter (Metric) 89 mm
Closure Type Screw Cap
Through the Closure Dispensing No