Learn More
Fisherbrand™ Unisex Lab Coats
Product Code. 15757287 Shop All Fisherbrand Products
Encompass
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Product Code. 15757287
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Knit wrist cuffs provide comfortable and secure fit
  • Made of 80/20 polycotton, 5 oz. performance-blend fabric
  • Precured durable press with soil-release treatment
  • One-piece, lined, notched lapel collar
  • 100% polyester-knit cuffs
  • Front gripper closures
  • One outside left breast pocket, two outside lower pockets Length: 41.5 in. (105cm) based on a size medium
  • White
Specifications
Size 2X-Large
Gender Unisex
Material Polyester, Cotton
Color White
Closure Type Snaps
Cuff Style Knit
Style Regular
No. of Pockets 3
Collar Type Notched lapel
Finish Pre-cure durable press
Item Description Gripper Snap
Length (English) 41.5 in.
Length (Metric) 105.4 cm
Construction Polyester, Combed Cotton
Show More Show Less
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.