Learn More
Fisherbrand™ Unisex Lab Coats
Product Code. 15737287 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Product Code. Size unitSize
15737287 Large Each
15717287 Small Each
15727287 Medium Each
15747287 X-Large Each
15757287 2X-Large Each
15767287 3X-Large Each
6 options
Product Code. 15737287
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Knit wrist cuffs provide comfortable and secure fit
  • Made of 80/20 polycotton, 5 oz. performance-blend fabric
  • Precured durable press with soil-release treatment
  • One-piece, lined, notched lapel collar
  • 100% polyester-knit cuffs
  • Front gripper closures
  • One outside left breast pocket, two outside lower pockets Length: 41.5 in. (105cm) based on a size medium
  • White
Specifications
Size Large
Gender Unisex
Material Polyester, Cotton
Color White
Closure Type Snaps
Cuff Style Knit
Style Regular
No. of Pockets 3
Collar Type Notched lapel
Finish Pre-cure durable press
Item Description Gripper Snap
Length (English) 41.5 in.
Length (Metric) 105.4 cm
Construction Polyester, Combed Cotton