Learn More
Fisherbrand™ Superfrost™ Plus Microscope Slides
Shop All Fisherbrand Products
Description
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
Same outstanding features as regular Superfrost slides with special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings.
- Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
- Chemical-resistant white marking surface
- Size: 25 × 75mm
Specifications
Specifications
| Length (Metric) | 75 mm |
| Width (Metric) | 25 mm |
| Frosted | ¾” SuperFrost Tab |
| Material | Glass |
| Type | Microscope Slide |
| Color | White |
| Corner Style | Regular |
| Quantity | 2 Packs of 72 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.