Learn More
Fisherbrand™ Square Labels
Shop All Fisherbrand Products
Click to view available options
Color:
Assorted
Blue
Green
Orange
Red
White
Yellow
Product Type:
Square Label
Quantity:
156/Pk.
52/Sheet
85 Labels/Sheet
Size (Metric):
0.64 x 3.17 cm
0.64 x 3.81 cm
0.64 x 3.81cm
1.91 x 4.29 cm
Unit Size:
20 sheets
25 sheets
Description
- Labels can be printed or written on and will not bleed, smudge or smear
- Engineered for use in laser printers
Specifications
Specifications
| Color | Green |
| Format | Sheet |
| Product Type | Square Label |
| Sterility | Non-sterile |
| Quantity | 85 Labels/Sheet |
| Size (English) | 26 x 1.25 in. |
| Size (Metric) | 0.64 x 3.17 cm |