Description
- Labels can be printed or written on and will not bleed, smudge or smear
- Engineered for use in laser printers
Specifications
Specifications
| Color | Orange |
| Format | Sheet |
| Product Type | Square Label |
| Sterility | Non-sterile |
| Labels per Sheet | 156 |
| Quantity | 156/Pk. |
| Size (English) | .25 x 1.5 in. |
| Size (Metric) | 0.64 x 3.81 cm |