Learn More
LevGo™ Inoculation Picks
Shop All Fisherbrand Products
Click to view available options
Height (Metric):
32.5 mm
32.51 mm
63.5 mm
152.4 mm
Size:
Large
Mini
Standard
Unit Size:
Pack of 1600
Pack of 2000
Pack of 500
Description
- Available in three lengths and two needle sizes to suit a wide range of application needs.
- Naturally biodegradable, made from 100% U.S.A grown white birch wood
- Manufactured in an FDA-registered factory and comply with FDA standards for food contact
- Available in semi or needle point styles for delicate operations
- Autoclavable
- Disposable
Specifications
Specifications
| Material | White Birch Wood |
| Size | Mini |
| Product Type | Toothpick |
| Quantity | 2000 Pk. |
| Autoclavable | Not Autoclavable |
| Height (Metric) | 32.5 mm |
| Height (English) | 1.28 in. |