Learn More
Fisherbrand™ Scientific Technical Pens
Product Code. 11724386 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Assorted
Black
Blue
Red
Quantity:
12 Pk.
6 Pk.
Width (Metric) Line:
0.2 mm
0.2, 0.4 mm
0.4 mm
Unit Size:
12 sets
Pack of 6
Product Code. Color Quantity Width (Metric) Line unitSize
11724386 Blue 6 Pk. 0.4 mm Pack of 6
11794376 Black 6 Pk. 0.4 mm Pack of 6
11784376 Black 6 Pk. 0.2 mm Pack of 6
10745512 Assorted 12 Pk. 0.2, 0.4 mm 12 sets
11704386 Red 6 Pk. 0.2 mm Pack of 6
11714386 Blue 6 Pk. 0.2 mm Pack of 6
6 options
Product Code. 11724386
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fast-drying ink in four colors
Fisherbrand™ Scientific Technical Pens produce extra-fine lines with accuracy—and without smearing. Special long-legged point is perfect for writing on notebooks, forms, graphs, charts, tables and recording paper. Fast-drying ink in four colors.
  • Ink is smooth flowing, fadeproof, and quick drying
  • Assortment includes red, blue, and special ultrablack for excellent reproduction on office copiers
  • Large ink reservoir lets you write for over 0.50 mile (800 m)
  • Cap design and color indicate point size and color of pen
  • Built-in super-tight cap seal keeps ink from drying out when pen is not in use
  • Choose from two line sizes: extra fine, 0.016 in. (0.4 mm), or super fine, 0.008 in. (0.2 mm)
  • Make precise lines with T-squares, rulers and flexible curves
Specifications
Product Type Technical Pen
Color Blue
Disposable Single-use
For Use With (Application) Used for record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams
Quantity 6 Pk.
Width (Metric) Line 0.4 mm