Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Product Code. 10340721
Change view
Click to view available options
Path Length:
1 mm
10 mm
Material:
Quartz Suprasil 300
Silica
Quantity:
2/Pk.
Ea.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Path Length Material Quantity Unit Size
10340721 10 mm Quartz Suprasil 300 Ea. Each
10258560 1 mm Quartz Suprasil 300 Ea. Each
12711675 10 mm Silica 2/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10340721 Supplier Hellma™ Supplier No. 1001046
  • ONLINE EXCLUSIVE PRICE
    2005.00 NOK
    1913.64 NOK / Pack of 1
    Save 91.36 NOK
    4% Off
Estimated Shipment: 04-12-2026
Includes
Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
  • Tried and tested over decades, used in countless applications
  • Utmost accuracy in terms of optical path length and parallelism
  • Very high temperature and chemical resistance due to thermal bonding of individual components
  • Unique surface flatness of the optical windows of 0.001mm
  • Function-optimized design with beveled edges and corners protects against the risk of damage caused by splitting and assists users in their daily work
Specifications
Type Macro Cell 100-QX
Description 2 windows
Capacity (Metric) 3500 μL
Material Quartz Suprasil 300
Width (Metric) Interior 9.5mm
Includes PTFE cover
Quantity Ea.
Path Length 10 mm
Wavelength Range 200 to 3500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm