missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps

Product Code. 11513802 Shop All Fisherbrand Products
Click to view available options
Unit Size:
Pack of 576
This item is not returnable. View return policy

Product Code. 11513802

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color White
Description Screw cap
Material HDPE
No. per Case 12/Pk.
Quantity 576/PK.
Sterility Sterile
Liner Material Polyvinyl
Neck Finish 24-400
Certifications/Compliance ISO 9001
Product Type Screw Cap
For Use With (Equipment) Bottle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.