Learn More
Fisherbrand™ Ostwald-Folin Pipets
Product Code. 17876237 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Accuracy:
±0.02 mL
Volume (Metric):
10 mL
Min. Flow Time:
5 sec.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Volume (Metric) Min. Flow Time Unit Size
17876237 ±0.02 mL 10 mL 5 sec. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17876237
Estimated Shipment: 07-01-2027
High-volume accuracy at a reasonable price.
  • Tapered bulbs
  • Borosilicate glass with permanent amber markings
  • Tuf-Temp™-treated to reduce chipping
  • Accuracy printed on each pipet
  • Accuracy equivalent to that of Class A volumetric
Order Info
Available as PersonaPet™ personalized pipets
Compliance
Calibrated To Deliver/Blow Out. Meet Fed. Spec. NNN-P-340B (or latest revision).
Specifications
Accuracy ±0.02 mL
Calibrated Yes
Certifications/Compliance NNN-P-340B
Material Borosilicate Glass
Min. Flow Time 5 sec.
Volume (English) 0.338 oz.
Color Yellow
Volume (Metric) 10 mL
Type Ostwald Folin Pipet