Learn More
Fisherbrand™ Digital Orbital Shaker
Product Code. 17811682 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Orbit:
19 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
17811682 19 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17811682
  • ONLINE EXCLUSIVE PRICE
    9690.00 NOK
    6997.56 NOK / Pack of 1
    Save 2692.44 NOK
    27% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Packed full of high-end design features, making it a versatile shaker for a vast range of protocols. Shaker has a speed range of 20-300 rpm in 1 rpm increments.
  • 5 Year Warranty
  • No-stop electronic motor feedback enables the unit to instantly adjust to weight load changes and unbalanced loads for truly reproducible results
  • Suitable for use in incubators (excl. CO2) and cold rooms within the stated temperature range
  • Soft-start and shut down feature at higher speeds reduces spill instances from full vessels
  • A high weight loading of 4.5 kg (10 lb.), with an overload protection cut out
  • Universal hi-lo platform and bungee cords allows you to create your own holding matrix for vessels of all shapes and sizes
  • Back lit display is easy to read.
  • Time and speed can be separately set
  • Very low start speed for sensitive sample agitation, with a high top end and good stability for more vigorous applications
  • Powered by 12 VDC, includes double insulated worldwide power adapter, 110 V to 240 V, supplied with 4 different plugs for power adapter
  • Long-life maintenance free motor and drive
Specifications
Orbit 19 mm
Load Bearing Capacity (Metric) 4.5 kg
Speed Control Accuracy 1 rpm increments
Speed Range 20 to 300 rpm
Electrical Requirements Universal
Voltage 110/240 V
Temperature Range (Metric) 2°C to 40°C
Wattage 18 W
Rotation Orbital
Holds Vessels of all shapes and sizes up to 4.5 kg (10 lb.) of load
Product Type Orbital platform shaker
Load Bearing Capacity (English) 10 lb.
Certifications/Compliance CE, CSA, RoHS3, UKCA, WEEE
Plug Type 4 interchangeable
Frequency 50/60 Hz
Material ABS plastic
Dimensions (LxW) Platform 11 x 11 in. (27.9 x 27.9 cm)
Dimensions (L x W x H) 12.4 x 7.8 x 12.2 in. (31.4 x 19.7 x 31 cm)
Height (English) 12.2 in.
Height (Metric) 31 cm
Length (English) 12.4 in.
Length (English) Platform 11 in.
Length (Metric) 31.4 cm
Length (Metric) Platform 27.9 cm
Width (English) 7.8 in.
Width (English) Platform 11 in.
Width (Metric) 19.7 cm
Width (Metric) Platform 27.9 cm
Shipping Weight (English) 27 lb.
Shipping Weight (Metric) 12.2 kg
Weight (English) 21.6 lb.
Weight (Metric) 9.8 kg
Model Digital Orbital Shaker
Display Digital display
For Use With (Equipment) Vessels of All Shapes and Sizes up to 4.5 kg. (10 lb.) of Load
Humidity Range 80% for temperatures up to 31°C decreasing
Includes Digital Orbital Shaker, Skid-Free Rubber Platform Mat, 8 Bungee Cord Set, Universal Hi-Lo Platform with Soft Rubber Edge Bumpers, 1 Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs
Operating Conditions 2°C to 40°C
Platform Style Square with silicone mat
Timer Range 0 to 74 hours 59 minutes (4499 minutes)