Learn More
Hellma Cylindrical Cells
Product Code. 10126690
Change view
Click to view available options
Path Length:
10 mm
50 mm
Unit Size:
Each
Product Code. Path Length unitSize
10126690 50 mm Each
12711705 10 mm Each
2 options
Product Code. 10126690
Brand: Hellma 1205040
Fit standard cell holders
Cylindrical cells have a filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
  • Uniform diameter of 22mm
  • Available with Suprasil™ quartz or near-UV glass windows
Specifications
Material Suprasil
Includes Suprasil quartz window; Two stopper
Path Length 50 mm