Learn More
Fisherbrand™ Cryogenic Vial Storage Boxes
Shop All Fisherbrand Products
Click to view available options
Holds:
1.2 to 2 mL Threaded Cryo Vials
5 mL Cryo Vials
Unit Size:
Pack of 24
Pack of 4
Description
Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials
- 81-Place white polycarbonate storage boxes
- Use in mechanical freezers and liquid nitrogen Dewars
- Cover is printed with a 9 x 9 numbered grid for inventory purposes
- Stackable
- Autoclavable
Specifications
Specifications
| Color | White |
| Material | Polycarbonate |
| Shape | Square |
| Type | Cryo Storage Box |
| For Use With (Application) | For ultra-low temperature storage of both internally and externally threaded cyrogenic vials |
| Holds | 5 mL Cryo Vials |
| Tube Array | 9 x 9 |
| Dimensions (L x W x H) | 5.25 x 5.25 x 3.75 in. (13.3 x 13.3 x 9.5 cm) |
| Length (English) | 5.25 in. |
| Length (Metric) | 13.3 cm |
| Show More |