Learn More
Bel-Art™ All-Metal Quick-Disconnects
Product Code. 10279681 Shop All SP Scienceware Products
Change view
Click to view available options
Diameter (English) Inner:
0.125 in.
Female, 0.25 in.
Male, 0.25 in.
For Use With:
3.2mm
Female, 6.4 mm
Male, 6.4 mm
Unit Size:
Each
Product Code. Diameter (English) Inner For Use With unitSize
10279681 0.125 in. 3.2mm Each
10107641 Male, 0.25 in. Male, 6.4 mm Each
10056381 Female, 0.25 in. Female, 6.4 mm Each
3 options
Product Code. 10279681
Brand: Bel-Art™ F197230802
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Inline hose connector with automatic shutoff on disconnect to prevent leakage
  • Chrome-plated brass hookups Compatible with most tubing
  • Operates at temperatures to 82°C (180°F), pressures to 250psi (1.7MPa) and vacuum to 30 in.Hg
Order Info
Order male and female connectors separately so tubing of the same or different size can be connected.
Specifications
Diameter (English) Inner 0.125 in.
Diameter (Metric) Inner 3.2 mm
Material Metal
For Use With 3.2mm
Maximum Pressure 250 psi
Product Type Quick Disconnect, Female Coupling