Learn More
Fisherbrand™ Cryogenic Dots, 0.5 in. Diameter
GREENER_CHOICE
Product Code. 17897221 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Assorted
Beige
Blue
Gold
Gray
Green
Lavender
Orange
Pink
Red
Silver
Violet
White
Yellow
Format:
Roll
Sheet
Unit Size:
20 sheets
25 sheets
Each
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Format unitSize
17897221 Blue Roll Each
17877201 Green Sheet 20 sheets
17887201 Orange Sheet 20 sheets
17897201 Red Sheet 20 sheets
17807211 Yellow Sheet 20 sheets
17867211 Assorted Sheet 25 sheets
17877211 Beige Roll Each
17807231 Gold Roll Each
17817231 Gray Roll Each
17827231 Green Roll Each
17837231 Lavender Roll Each
17847231 Orange Roll Each
17857231 Pink Roll Each
17867231 Red Roll Each
17897231 Yellow Roll Each
17887231 Violet Roll Each
17877231 Silver Roll Each
17817241 White Roll Each
17807241 Assorted Roll Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 17897221
Add to basket
Add to basket
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.
Specifications
Color Blue
Format Roll
Product Type Dot Label
Sterility Non-sterile
Includes Roll of 1000 dots
Diameter (English) 0.5 in.
Diameter (Metric) 12.7 mm