Learn More
Fisherbrand™ Spacers for Owl™ P10DS Dual Gel System
Product Code. 11953065 Shop All Thermo Scientific Products
Change view
Click to view available options
Thickness (Metric):
0.08 cm
0.15 cm
Height (Metric):
0.08 cm
0.15 cm
Unit Size:
Pack of 2
Product Code. Thickness (Metric) Height (Metric) unitSize
11953065 0.15 cm 0.15 cm Pack of 2
11943065 0.08 cm 0.08 cm Pack of 2
2 options
Product Code. 11953065
Brand: Fisherbrand™ P2SD
Only null left
Add to basket
Add to basket
Spacers for Owl™ P10DS Dual Gel System
  • For use with Owl P10DS dual gel system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P10DS Dual Gel System
Type Spacer Set
Gel Dimensions 20 x 20 cm
Thickness (English) 0.05 in.
Thickness (Metric) 0.15 cm
Length (English) Footprint 9.84 in.
Length (English) Gel 7.8 in.
Length (Metric) Footprint 25 cm
Length (Metric) Gel 20 cm
Width (English) Footprint 5.11 in.
Width (English) Gel 7.8 in.
Width (Metric) 0.1 cm
Width (Metric) Footprint 13 cm
Width (Metric) Gel 20 cm
Height (English) 0.05 in.
Height (English) Footprint 9.05 in.
Height (Metric) 0.15 cm
Height (Metric) Footprint 23 cm