Learn More
Fisherbrand™ Spacers for FB-GC10-1 Vertical Gel Caster
Product Code. 11993065
Change view
Click to view available options
Thickness (Metric):
0.8 cm
1.5 cm
Height (Metric):
0.008 cm
0.15 cm
Unit Size:
Pack of 2
Product Code. Thickness (Metric) Height (Metric) unitSize
11993065 0.8 cm 0.008 cm Pack of 2
11903075 1.5 cm 0.15 cm Pack of 2
2 options
Product Code. 11993065
Brand: Fisherbrand™ P7SC
Only null left
Add to basket
Add to basket
Spacers for use with FB-GC10-1 Vertical Gel Caster.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P7-CST Multiple Gradient Gel Caster
Type Spacer Set
Gel Dimensions 10 x 10 cm
Includes Two side spacers and one bottom spacer
Thickness (English) 0.03 in.
Thickness (Metric) 0.8 cm
Length (English) Gel 3.9 in.
Length (Metric) Gel 10 cm
Width (English) Footprint 5.9 in.
Width (English) Gel 3.9 in.
Width (Metric) 0.05 cm
Width (Metric) Footprint 15 cm
Width (Metric) Gel 10 cm
Height (English) 0.003 in.
Height (Metric) 0.008 cm