Learn More
Fisherbrand™ Sequencing Gel Caster
Product Code. 11957515 Shop All Thermo Scientific Products
Change view
Click to view available options
:
Gel size: 20 to 42 x 48cm
Product Code.
11957515 Gel size: 20 to 42 x 48cm
1 options
Product Code. 11957515
Brand: Fisherbrand™ SGC1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Adjustable casting platform, width adjustment knobs, and 12 binder clamps
Bubble-free sequencing gels in less than three minutes
  • Simple, innovative, and quick method for casting
  • No taping or special assembly is required
  • System is adjustable—accommodates glass plates from 20 to 42cm wide and 48cm long
  • Horizontal plate sliding, capillary action drawing gel between plates, and surface tension keep gel from leaking
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 12.2 cm
Length (English) Gel 7.8 to 16.5 in.
Length (Metric) 71 cm
Length (Metric) Gel 20 to 42 cm
Product Type Sequencing Gel Caster
Width (English) Gel 25.7 in.
Width (Metric) 21 cm