Learn More
Fisherbrand™ Preparative/Marker Comb for Owl™ P2-CST Multiple Gradient Gel Caster
Product Code. 11933065
Click to view available options
Unit Size:
Each
Product Code. 11933065
Brand: Fisherbrand™ P2PREP
Only null left
Add to basket
Add to basket
Preparative/Marker Comb for Owl™ P2-CST Multiple Gradient Gel Caster
  • For use with Owl P2-CST Multiple Gradient Gel Caster
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P2-CST Multiple Gradient Gel Caster
Type Preparative/Marker Comb
Gel Dimensions 10 x 20 cm
Teeth Count 2
Thickness (Metric) Teeth 0.15 cm
Volume (Metric) Well 448/155 μL
Length (English) Gel 7.8 in.
Length (Metric) Gel 20 cm
Width (English) Gel 7.8 in.
Width (English) Teeth 5.8/0.18 in.
Width (Metric) Gel 20 cm
Width (Metric) Teeth 14.81/0.47 cm