Learn More
Fisherbrand™ Pouches for FB-GC10-1 Vertical Gel Caster
Click to view available options
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
Description
- Dimensions: 10L × 10cmW
Specifications
Specifications
| For Use With (Equipment) | Owl JGC-4 Gel Casting System |
| Type | Gel Pouch |
| Quantity | 100/Pk. |
| Length (Metric) | 10 cm |
| Width (Metric) | 10 cm |