Learn More
Fisherbrand™ Hybridization Tubes
Shop All Fisherbrand Products
Click to view available options
Length (English):
3.93 in.
5.90 in.
11.81 in.
Diameter (English) Inner:
1.37 in.
2.75 in.
Unit Size:
Each
Pack of 2
Description
Fits FisherBiotech™ Hybridization Incubator rotisseries and most rotisseries that accept standard 35 or 70mm diameter tubes.
- Thick-walled borosilicate glass
- Includes polypropylene cap
Specifications
Specifications
| Closure Material | Polypropylene |
| Diameter (English) Inner | 2.75 in. |
| Diameter (Metric) Inner | 70 mm |
| Diameter (Metric) Outer | 70 mm |
| For Use With (Equipment) | FisherBiotech hybridization incubator rotisseries |
| Length (English) | 11.81 in. |
| Closure Type | With Cap |
| Length (Metric) | 300 mm |
| Material | Borosilicate Glass |
| Product Type | Hybridization Tube |