Learn More
Fisherbrand™ DNA Sequencing Systems: Wedge Spacer Set for Discontinued Model FB-SEQ-2045
Product Code. 12404727
Change view
Click to view available options
Quantity:
2/Pk.
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12404727 2/Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12404727 Supplier Fisherbrand™ Supplier No. S2SSAW
  • ONLINE EXCLUSIVE PRICE
    1375.00 NOK
    1333.66 NOK / Pack of 2
    Save 41.34 NOK
    3% Off
Estimated Shipment: 07-12-2026
DNA Sequencing Systems Wedge Spacer Set for discontinued model FB-SEQ-2045
For use with discontinued model FB-SEQ-2045
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Sequencing gel casting systems
Height (Metric) 0.04 cm
Product Type Wedge Spacer Set
Quantity 2/Pk.
Thickness (Metric) Teeth 0.4 to 0.8 mm
Width (Metric) 1 cm