Learn More
Fisherbrand™ 20-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
Product Code. 11973055 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric) Well:
97 μL
182 μL
Thickness (Metric) Teeth:
0.08 cm
0.15 cm
Unit Size:
Each
Product Code. 11973055
Brand: Fisherbrand™ P220C
Only null left
Add to basket
Add to basket
20-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
  • For use with Owl P2-CST Multiple Gradient Gel Caster
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P2-CST Multiple Gradient Gel Caster
Type Comb
Gel Dimensions 20 x 20 cm
Teeth Count 20
Thickness (Metric) Teeth 0.08 cm
Volume (Metric) Well 97 μL
Length (English) Footprint 9.84 in.
Length (English) Gel 7.8 in.
Length (Metric) Footprint 25 cm
Length (Metric) Gel 20 cm
Width (English) Gel 7.8 in.
Width (English) Teeth 0.21 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 20 cm
Width (Metric) Teeth 0.55 cm
Height (English) Footprint 9.05 in.
Height (Metric) Footprint 23 cm