Learn More
Fisherbrand™ 10-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
Product Code. 11933055 Shop All Thermo Scientific Products
Change view
Click to view available options
Thickness (English):
0.8mm
1.5mm
Unit Size:
Each
Product Code. Thickness (English) unitSize
11933055 1.5mm Each
11923055 0.8mm Each
2 options
Product Code. 11933055
Brand: Fisherbrand™ P210D
Only null left
Add to basket
Add to basket
10-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
  • For use with Owl P2-CST Multiple Gradient Gel Caster
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P2-CST Multiple Gradient Gel Caster
Type Comb
Gel Dimensions 20 x 13 cm
Teeth Count 10
Thickness (English) 1.5mm
Thickness (Metric) Teeth 0.15 cm
Volume (Metric) Well 449 μL
Length (English) Footprint 5.11 in.
Length (English) Gel 7.8 in.
Length (Metric) Footprint 13 cm
Length (Metric) Gel 20 cm
Width (English) Gel 7.8 in.
Width (English) Teeth 0.53 in.
Width (Metric) Footprint 25 cm
Width (Metric) Gel 20 cm
Width (Metric) Teeth 1.36 cm
Height (English) Footprint 9.05 in.
Height (Metric) Footprint 23 cm