Learn More
Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Wash Heads
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer
Wellwash Versa Microplate Strip Washer
Type:
Blank Wash Head
Wash Head
Unit Size:
Each
Holds:
2 x 8
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Applications:
ELISA-based assays, cell washing
Specifications
Specifications
| For Use With (Application) | ELISA-based assays, cell washing |
| Type | Blank Wash Head |
| For Use With (Equipment) | Wellwash Versa Microplate Strip Washer |