Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Product Code. 11797996 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
9 L
Type:
Rinse Bottle
Wash Bottle
Wash Bottle A
Wash Bottle B
Waste Bottle
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Type unitSize
11797996 4 L Wash Bottle A Each
11708006 4 L Wash Bottle B Each
11737996 4 L Waste Bottle Each
11718006 4 L Rinse Bottle Each
11758006 9 L Waste Bottle Each
11777996 2 L Rinse Bottle Each
11767996 2 L Wash Bottle B Each
11727996 2 L Waste Bottle Each
11747996 2 L Wash Bottle Each
11757996 2 L Wash Bottle A Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11797996 Supplier Thermo Scientific™ Supplier No. N10811
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
TRUSTED_SUSTAINABILITY
Specifications
Includes Cap with tubing and liquid level sensors
Capacity (English) 135.25 oz.
Capacity (Metric) 4 L
Shape Round
Type Wash Bottle A
For Use With (Equipment) Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer