Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Product Code. 11718006 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
2 L
4 L
9 L
Type:
Rinse Bottle
Wash Bottle
Wash Bottle A
Wash Bottle B
Waste Bottle
Unit Size:
Each
Product Code. 11718006
Brand: Thermo Scientific™ N10813
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 135.25 oz.
Capacity (Metric) 4 L
Shape Round
Type Rinse Bottle
For Use With (Equipment) Wellwash Versa Microplate Strip Washer