Learn More
Fisherbrand™ Traceable™ Pure H2O Tester
Shop All Fisherbrand Products
Click to view available options
Range:
0.1 to 20000μS
Unit Size:
Each
Description
Includes
Probe with 40 in. cable; a Traceable Certificate to indicate instrument traceability to NIST from an ISO 17025 calibration laboratory; carrying case; instructions and 9V battery.
Accurate readings in an instant. Use to test water from stills, demineralizers, deionizers, and reverse osmosis equipment.
- Simple operation eliminates errors
- Internal, solid-state thermometer automatically references readings to international standard of 25°C (77°F)
- Accuracy is ±0.4% of full scale
- Range is 0.1 to 20,000 micromhos (from pure to raw water)
- Comes ready to operate
Compliance
Meets all lab certification requirements for pure water analysis for CAP, ASTM™, NCCLS, USP, and ACS.
Specifications
Specifications
| Certifications/Compliance | CAP, ASTM, NCCLS, USP, and ACS |
| For Use With (Application) | Single purpose unit designed for labs required to maintain a periodic check of water purity |
| For Use With (Equipment) | Demineralizers, deionizers, reverse osmosis equipment |
| Includes | Probe with 40 in. cable, traceable™ certificate to indicate instrument traceability to NIST from an ISO 17025 calibration laboratory, carrying case, instructions and 9V battery |
| Range | 0.1 to 20000μS |
| Type | Traceable™ Pure H2O Tester |