Learn More
Fisherbrand™ Tip Horn for Model 120 Sonic Dismembrator
Product Code. 12357338 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
0.3 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12357338 0.3 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12357338
  • ONLINE EXCLUSIVE PRICE
    15765.00 NOK
    14906.25 NOK / Pack of 1
    Save 858.75 NOK
    5% Off
Estimated Shipment: 03-12-2026
Tip Horn for Model 120 Sonic Dismembrator
This Fisher Scientific Tip Horn is designed for use with the Model 120 Sonic Dismembrator
  • 8-channel tip horn
  • For use with Model 120 Sonic Dismembrator
Specifications
Product Type Tip Horn
Diameter (English) 0.12 in.
Diameter (Metric) 0.3 cm
For Use With (Equipment) Model 120 sonic dismembrator