Description
Fisher Scientific™ Technical Pens are ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams. 6 per pack.
Available in multiple colors and widths.
Specifications
Specifications
| Tip Style | Fine |
| Product Type | Technical Pen |
| Color | Green |
| For Use With (Application) | Ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams |
| Quantity | 6 Pk. |
| Width (Metric) Line | 0.4 mm |