Learn More
Fisherbrand™ Stackable Microplate Holder for Microplate Vortex Mixers
Shop All Fisherbrand Products
Click to view available options
No. of Positions:
8
Unit Size:
Each
Description
- To maximize capacity of Heavy-Duty Vortex Mixer to eight microplates by stacking the tray on top of four plate holder
Specifications
Specifications
| Product Type | Stackable microplate holder |
| For Use With (Equipment) | Microplate vortex mixers 02-216-108 and 02-216-109 |
| No. of Positions | 8 |