Learn More
Fisherbrand™ Model 705 Sonic Dismembrator Accessory
Product Code. 15310382 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
4-tip Horn
Replacement Tip
Unit Size:
Each
Product Code. Product Type unitSize
15310382 4-tip Horn Each
15320382 Replacement Tip Each
2 options
Product Code. 15310382
Only null left
Add to basket
Add to basket
Requires
Cat. No. FB4659
4 Tip horn for use with 500 or 700 watt sonicator systems; replacement tip also available.
  • For processing 0.5 to 15ml volumes
  • Process 4 samples at one time
Specifications
Product Type 4-tip Horn
For Use With (Application) Lyse cells, shear DNA, dissolve solids
For Use With (Equipment) FB705 Sonic Dismembrator and Q700 Sonicator