Learn More
Fisherbrand™ Isotemp™ Advanced Stirrer
Shop All Fisherbrand Products
Click to view available options
Electrical Requirements:
230V 50/60Hz
Plug Type:
EU
Top Plate Surface Area (Metric):
18.4 x 18.4 cm
Unit Size:
Each
Description
Intuitive design
- Top plate choices in ceramic or aluminum
- Raised display design protects electronics from spills
- Ability to set programs (ramp/dwell) as well as save settings
- Includes stir bar included
- Optional splash guard protection shield
Advanced technology
- StirTrac features innovative engineering for slow-speed stirring, consistent speed control, and stronger magnetic coupling
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Warranty
Three years
Specifications
Specifications
| Speed | 50 to 1500 rpm |
| Top Plate Material | Ceramic |
| Capacity | 4 L |
| Voltage | 230 V |
| Dimensions (L x W x H) | 13 x 8.2 x 3.8 in. (33 x 20.8 x 9.7 cm) |
| Top Plate Surface Area (English) | 7.25 x 7.25 in. |
| Top Plate Surface Area (Metric) | 18.4 x 18.4 cm |
| Electrical Requirements | 230V 50/60Hz |
| Frequency | 50/60 Hz |
| Plug Type | EU |
| Show More |